TWS119 10mM in DMSO, EZSolution™, sterile-filtered
Catalog # 76303-446
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic vial
- Size:5 mg
- Cat. No.:76303-446
Specifications
About this item
:
:318.34 g/mol
Storage Temperature:Freezer
UN:1993
ADR:CBL,III
CAS Number:601514-19-6
Formula:C₁₈H₁₄N₄O₂
MDL Number:MFCD09037535