Modified Medium 199, MP Biomedical, LLC.
Catalog # IC091023122
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Powder
- Description:Modified Medium 199
- Size:1× 10 L
- Storage Temperature:0...5 °C
- Cat. No.:IC091023122
Specifications
About this item
Culture Media Medium with Hanks' Salts with Glutamine and without Sodium Bicarbonate.