MicroMaxNS Disposable Protective Coveralls with Open Wirsts and Ankles, Zipper Closure, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
These disposable protective coveralls are made from lightweight microporous fabric.
- Microporous film laminated to polypropylene substrate
- Serged seams
- Blood-borne pathogen/viral barrier tested
MicroMax NS coverall offers the protection against dirt, dust, grease, grime and light chemical splash.
Open wrists and ankles for increased comfort.
Specifications
- Catalog No:
- 76309-054
- 76309-056
- 76309-060
- 76309-062
- Fabric:
- PP + laminated film
- PP + laminated film
- PP + laminated film
- PP + laminated film
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- Seam type:
- Serged
- Serged
- Serged
- Serged