StemBoost™ Neural Stem Cell Reprogramming Cocktail (100X), Biovision
Supplier: Avantor
Certificates
About this item
A neural stem cell reprogramming cocktail
- Appearance: Liquid
- Formulation: In DMSO
- Handling: Protect from air and light
- Storage Conditions: -20°C
- Shipping Conditions: Gel Pack
StemBoost™ Neural Stem Cell Reprogramming Cocktail is a convenient, sterile-filtered cocktail solution (100X, in DMSO) containing eight (8) small molecules useful for reprogramming of fibroblasts into neural stem cells. The eight compounds are: CHIR99021 (Cat. No. 1677), LDN193189 (Cat. No. 1995), A83-01 (Cat. No. 1725), Tranylcypromine hemisulfate (Cat. No. 1816), RG 108 (Cat. No. 1679), Retinoic acid (Cat. No. 2218), SMER28 (Cat. No. 1856), and Hh Ag 1.5 (Cat. No. B2160).
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together