Order Entry
United States
ContactUsLinkComponent
1X Hanks' Balanced Salt Solution (Modified) without Phenol Red, MP Biomedical
1X Hanks' Balanced Salt Solution (Modified) without Phenol Red, MP Biomedical
  IC091810254
 :  
undefined
1X Hanks' Balanced Salt Solution (Modified) without Phenol Red, MP Biomedical
  IC091810254
 :  091810254
 :  

 

  • Description:
    1X Hanks' balanced salt solution (modified) without phenol red
  • Size:
    500 mL
  • Sterility:
    Sterile
  • pH Range:
    6.9 - 7.4
  • Storage Temperature:
    Room temperature (15…30 °C).
  • Physical Form:
    Clear, colorless solution
  • Cat. No.:
    IC091810254

 

 

Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.

  • Stem cells growth
  • Growth of cells at low densities