1X Hanks' Balanced Salt Solution, Modified, MP Biomedical, LLC.
Catalog # IC091810054
Supplier: MP Biomedicals
CAS Number:
Specifications
- Description:1X Hanks' Balanced Salt Solution, Modified
- Size:500 mL
- pH Range:6.9 - 7.2
- Storage Temperature:Room Temperature (15…30 °C).
- Physical Form:Red; Clear Solution
- Cat. No.:IC091810054