About this item
BioFX® Tris Buffered Saline/Casein Block and Diluent-10X Concentrate minimizes non-specific interactions of the secondary reactants with each other and with the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins.
- 2 year shelf life
- Minimizes non-specific binding
- Ready to use formulation
The solution is ready for dilution for specific application requirements.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Shelf Life:2 years
- Storage Buffer:Tris buffered saline (TBS)
- Storage Temperature:2…8 °C