ContactUsLinkComponent
Vessel Caps with TFM Septa, CEM
Vessel Caps with TFM Septa, CEM
Catalog # MSPP-909355
Supplier:  CEM CORPORATION MS
Vessel Caps with TFM Septa, CEM
Catalog # MSPP-909355
Supplier:  CEM CORPORATION MS
Supplier Number:  909355
Specifications
  • Description:
    Vessel caps with TFM septa
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-909355
  • Fits:
    35/80 ml vials
  • Supplier no.:
    909355
Specifications
About this item
These caps are compatible with Discover SP-D and Discover SP-D 80 for microwave digestion.