Order Entry
United States
ContactUsLinkComponent
Electrodes Storage Solution, Metrohm
Electrodes Storage Solution, Metrohm
Catalog # MSPP-62323000
Supplier:  METROHM USA MS
CAS Number:  
Electrodes Storage Solution, Metrohm
Catalog # MSPP-62323000
Supplier:  METROHM USA MS
Supplier Number:  6.2323.000
CAS Number:  

Specifications

  • Description:
    Storage Solution
  • Size:
    250 ml
  • Cat. No.:
    MSPP-62323000

Specifications

About this item

Storage solution for all combined pH glass electrodes with reference electrolyte c(KCl) = 3 mol/l.