NEW AGE® 9128 Series Nitrile Examination Gloves, Tronex
: Tronex
Innovative nitrile material formulation fosters exceptional 'fit & feel' properties for applications with shorter durations.
- Specialized formulation for lightness of fit and superior tactile sensitivity
- Not made with natural rubber latex; powder-free; non-sterile
- Fingertip textured for improved grip
- Ambidextrous with beaded cuffs
Offers excellent barrier protection from bloodborne pathogens, as well as resistance to chemotherapy drugs and other chemicals.
: FDA 510(k) approved for medical use.
ASTM D6319; ASTM D6978; ASTM F1671
Manufacturing certified ISO 9001 for Quality Management Systems, as well as ISO 13485 for Quality Management Systems for Medical Devices.
ASTM D6319; ASTM D6978; ASTM F1671
Manufacturing certified ISO 9001 for Quality Management Systems, as well as ISO 13485 for Quality Management Systems for Medical Devices.
: 250 gloves/box, 10 boxes/case, total: 2500 gloves.
- Catalog No:
- 76018-958
- 76018-950
- 76018-952
- 76018-954
- 76018-956
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Fingertips textured
- Fingertips textured
- Fingertips textured
- Fingertips textured
- Fingertips textured
- Cuff Style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- FDA Regulated:
- +
- +
- +
- +
- +
- ASTM No.:
- ASTM D6319; ASTM D6978; ASTM F1671
- ASTM D6319; ASTM D6978; ASTM F1671
- ASTM D6319; ASTM D6978; ASTM F1671
- ASTM D6319; ASTM D6978; ASTM F1671
- ASTM D6319; ASTM D6978; ASTM F1671
Frequently Bought Together