Ultra Sensitive Mouse Insulin ELISA Kit, Crystal Chem
Catalog # MSPP-90080
Supplier: CRYSTAL CHEM INC MS
CAS Number:
Specifications
- Primary antibody reactivity:Mouse
- Description:Ultra Sensitive Mouse Insulin ELISA Kit
- Size:1 kit
- Sample Type:Serum, Plasma, Cell Culture, or Fluid
- Storage Temperature:2...8 °C
- Sample Volume:5 µl
- Sensitivity:0.05 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-90080
- Supplier no.:90080
- No. of tests:96 wells
Specifications
About this item
The insulin mouse Ultra sensitive ELISA is used to quantify levels of mouse insulin in serum, plasma, cell culture, and fluid using only a 5 µl sample volume.
- Kits use only 5 µl sample
- Dynamic range of 0.1 to 64 ng/ml
- High sensitivity (50 pg/ml) using 5 µl sample
- Specific and fast
- Recovery: When mouse insulin was spiked to mouse serum, the recovery of mouse insulin was 100% ±15%
Ultra sensitive mouse insulin ELISA kit have the low range assay of 0.1 to 6.4 ng/ml, wide range assay of 0.1 to 12.8 ng/ml and high range assay of 1 to 64 ng/ml.