Order Entry
United States
ContactUsLinkComponent
Iscove's Modified Dulbecco's Medium (IMDM), ATCC® 30-2005™
Iscove's Modified Dulbecco's Medium (IMDM), ATCC® 30-2005™
Catalog # MSPP-302005
Supplier:  0000013421
CAS Number:  
Iscove's Modified Dulbecco's Medium (IMDM), ATCC® 30-2005™
Catalog # MSPP-302005
Supplier:  0000013421
Supplier Number:  30-2005
CAS Number:  

Specifications

  • Description:
    Iscove's modified dulbecco's medium (IMDM)
  • Size:
    500 ml
  • Storage Temperature:
    2...8 °C
  • Cat. No.:
    MSPP-302005
  • Supplier no.:
    30-2005

Specifications

About this item

Iscove's modified dulbecco's medium (IMDM) contains 4 mM L-glutamine, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate.

  • pH: 7.0 to 7.4
  • Osmolality: 235 to 309 mOsm/kg
  • Sterility testing: Pass
  • Endotoxin: ≤0.5 EU/ml
  • Mycoplasma: Pass
  • Growth promotion: Pass

Iscove's modified dulbecco's medium (IMDM) reduced level of sodium bicarbonate (NaHCO₃, 1.5 g/L) is intended for use in 5% CO₂ in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO₂.