ContactUsLinkComponent
Weller® Precision Tweezer A Standard Model, Apex Tool Group
Weller® Precision Tweezer A Standard Model, Apex Tool Group
Catalog # 76238-630
Supplier:  Cooper Tools
Weller® Precision Tweezer A Standard Model, Apex Tool Group
Catalog # 76238-630
Supplier:  Cooper Tools
Supplier Number:  EROP3CSA
Specifications
  • Length:
    110 mm
  • Material:
    Special Stainless Steel
  • Cat. No.:
    76238-630
Specifications
About this item
Designed for delicate work.
  • Suitable for delicate standard applications
  • Precision work on small components or wires
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heatresistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers.