ContactUsLinkComponent
Weller® Precision Curved Tweezer with Pointed Tips, Apex Tool Group
Weller® Precision Curved Tweezer with Pointed Tips, Apex Tool Group
Catalog # 76238-622
Supplier:  Cooper Tools
Weller® Precision Curved Tweezer with Pointed Tips, Apex Tool Group
Catalog # 76238-622
Supplier:  Cooper Tools
Supplier Number:  7SA
Specifications
  • Length:
    120 mm
  • Material:
    Special Stainless Steel
  • Environmentally Preferable:
  • Cat. No.:
    76238-622
  • Tip style:
    Curved
Specifications
About this item
Bent shape facilitates access to confined spaces.
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heat resistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers. For applications in biology, medicine, laboratory technology, and microelectronics.