About this item
ZymeFree™ cell dissociation reagent in Dulbecco's phosphate buffered saline can be used for gentle removal of adherent cells from each other and from the surface of a culture vessel.
- ZymeFree™ works as quick as trypsin on most cell lines
- Osmolality in mOsm/Kg H₂O: 280.00 to 320.00
- Endotoxin content: NMT 5 EU/ml
This enyme free formulation helps in preserving the structural and functional integrity of the cell surface proteins. It also helps in avoiding the cytotoxic effects sometimes associated with the chelating agents. This product is very useful for studies that require intact cell surface proteins such as ligand binding, flow cytometry and immunohistochemistry. It can also be used as a trypsin alternative in serum free culture work.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: