Flow Injection System Supplies, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
Several of modular building block type mixing manifold blocks can be connected in series for enhanced mixing capabilities.
- Modular building blocks are made of chemically-resistant plastic
Modular building block type gas/liquid separator: An exchangeable PTFE membrane in the screw cap prevents liquid from being carried into the quartz cell when working with strong foaming samples.
Delivery: Complete modular mixing/separation assembly includes two mixing manifolds with tubing adapters, gas liquid separator with PTFE membrane, 5 spare PTFE membranes, PTFE tubes (110 and 300 mm). Modular building block type mixing manifold includes mixing channel and 3 connections, 2 inlet and 1 outlet.
Frequently Bought Together