ContactUsLinkComponent
Universal Vial Caps, Ab Sciex
Universal Vial Caps, Ab Sciex
Catalog # MSPP-A62250
Supplier:  AB SCIEX LLC MS
Universal Vial Caps, Ab Sciex
Catalog # MSPP-A62250
Supplier:  AB SCIEX LLC MS
Supplier Number:  A62250
Specifications
  • Description:
    Universal Vial Caps
  • Cat. No.:
    MSPP-A62250
  • Supplier no.:
    A62250
Specifications
About this item
Universal caps are used in combination with universal vials.
Universal vial caps are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.