Order Entry
United States
ContactUsLinkComponent
Kimax® Borosilicate Pertri Dish Covers, DWK Life Sciences
Kimax® Borosilicate Pertri Dish Covers, DWK Life Sciences
Catalog # 89001-250
undefined
Kimax® Borosilicate Pertri Dish Covers, DWK Life Sciences
Catalog # 89001-250
Supplier Number:  23062-10010

Specifications

  • Ø:
    100 mm
  • Description:
    Petri Dish Replacement Cover
  • Height:
    10 mm (0.39")
  • Environmentally Preferable:
  • Cat. No.:
    89001-250
  • Shape:
    Round
  • Supplier no.:
    23062-10010

Specifications

About this item

Replacement glass covers can be used with dishes for culturing bacteria and other general laboratory applications.

  • Covers remain clear after repeated use in wet or dry sterilization cycles
  • Reinforced beaded edges resist mechanical breakage
  • Combination of different colors and wording permits quick and easy identification
  • Manufactured from 33 expansion, low extractable borosilicate glass

Covers have a fused-on bright red ceramic enamel marking and inscription.

Certifications: Designed to meet Federal Specification NNN-D-1478. Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.