Order Entry
United States
ContactUsLinkComponent
 
UVP PCR Cabinet and Workstations, Analytik Jena
 
undefined
UVP PCR Cabinet and Workstations, Analytik Jena

 

  • Catalog No:
  • 76302-866
  • 76303-016
  • 76420-640
  • 76420-638
  • No. of Outlets:
  • Two built-in power outlets
  • Two built-in power outlets
  • Two built-in power outlets
  • Two built-in power outlets
  • Light Source:
  • 254 nm (Pre-filter chamber, work area, UV/air recirculator) and overhead white LED
  • 254 nm (Pre-filter chamber, work area, UV/air recirculator) and overhead white LED
  • 254 nm (Pre-filter chamber, work area, UV/air recirculator) and overhead white LED
  • 254 nm (Pre-filter chamber, work area, UV/air recirculator) and overhead white LED
  • Safety:
  • UV shut-off switches in all chambers and red LED ambient light to indicate UV on
  • UV shut-off switches in all chambers and red LED ambient light to indicate UV on
  • UV shut-off switches in all chambers and red LED ambient light to indicate UV on
  • UV shut-off switches in all chambers and red LED ambient light to indicate UV on
  • W×D×H:
  • 826×616×991 mm (32.5×24.25×39")
  • 826×616×991 mm (32.5×24.25×39")
  • 686×508×889 mm (27×20×35")
  • 686×508×889 mm (27×20×35")

 

Frequently Bought Together