To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- L×W:82.6×142.9 mm
- Thickness:114 µm
- Cat. No.:101414-322
- Supplier no.:AC1201-02
Specifications
About this item
A 114 µm thick, hydrophobic, porous film with medical-grade adhesive for tissue culture plates, bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth.
AeraSeal films minimize cross-contamination, spillage, and evaporation. They allow uniform air and CO2 exchange for all wells, unlike the plate lids which favor exchange for wells near plate edges.