Replaceable Fiber Tipped Tweezers, Lindstrom Precision Tools
Supplier: J.H. Williams Tool Group (Lindstrom)
Certificates
About this item
Polished stainless steel body with replaceable plastic tips. ESD carbon fiber tips do not leave metal marks and grant a contamination free performance.
- Easy tip replacement system
- Replacement tips include two tips and two screws in a plastic bag
Frequently Bought Together