ContactUsLinkComponent
Test Tubes, Charles River Endosafe MS
Test Tubes, Charles River Endosafe MS
  MSPP-T300
Test Tubes, Charles River Endosafe MS
  MSPP-T300
 :  T300
  • Ø:
    13 mm
  • Description:
    Tubes wrapped in aluminium foil
  • Length:
    100 mm
  • Material:
    Borosilicate glass
  • Cat. No.:
    MSPP-T300
  • Packaging:
    50/Pack
  • Supplier no.:
    T300