Saf-T-Pak® Category A Frozen Insulated Shipping System
Catalog # MSPP-STP310
Supplier: SAF-T-PAK INC MS
Specifications
- Description:Refrigerated infectious shipper, insulated, stp 310
- Environmentally Preferable:
- Cat. No.:MSPP-STP310
- Supplier no.:STP-310
Specifications
About this item
Ideal for use with Saf-T-Temp™ products or Dry ice for thermal control.
- Insulated chest lid
- Leakproof polybag
- Insulated chest inside a fibreboard box
Category B / Empty Package Conversion Panel for maximum value.
Approximate Capacity*:
Max. Secondary Pressure Receptacle(s) (STP-710): 3
Max. 2mL Tubes: 243
Max. 10mL Tubes: 32
Max. 14mL Tubes: 32
Max. 50mL Centrifuge Tubes: 8
Max. 500mL Blood Bag(s): 2
Max. 90mL Petri Dishes: 3
Delivery: Contains reusable 4G outer box, polystyrene cooler and lid, STP 111 inner box, empty package cover, two STP 710, secondary containers, Two 250 ml absorbent, strips, hazard and handling labels, 3 copies shipper’s declaration form, instruction manual, two 12×12" bubble wrap.