Microloader™ Tips, Eppendorf
Catalog # 89009-310
Supplier: Avantor
Product available on GSA Advantage®
Specifications
- Volume:0.5 - 20 µl
- Length:100 mm
- Packed:96/Rack, 2 Racks/case
- Cat. No.:89009-310
- Tip style:Microloader Tips
- For use with:Femtotips™
- Supplier no.:930001007
Specifications
About this item
Microloader™ tip for filling Femtotips and other glass microcapillaries (for research use only).
- Microloader tips and rack autoclavable
Extremely long, fine and flexible tip for filling of microcapillaries for microinjection and where additional reach is needed. Highest precision and accuracy when used with Eppendorf pipettes for 0.5 to 10 µl (gray control button).