Eyewear Cleaning Wipes Box, HexArmor
Catalog # 76207-856
Supplier: HexArmor
Specifications
- Description:Eyewear Cleaning Wipes Box
- Cat. No.:76207-856
- Packaging:100/Box
Specifications
About this item
Individually sealed, anti-static cleaning wipes for all lens types.
- Pre-moistened and silicone-free
- Packaged 100 wipes/box
Proprietary coatings are chemically formulated to perform differently than any other coating on the market. Therefore, using standard, non-HexArmor, alcohol-based cleaner on them can cause damage.
For this reason, specific cleaning supplies are available to ensure the TruShield® coatings continuously perform to the highest standard. Using any anti-fog wipes, alcohol-based cleaners, or any other non-HexArmor product on TruShield® lenses can cause damage.