Covered and Uncovered Flat-Bed Slide Warmers
Supplier: C & A Scientific
Certificates
About this item
These flat topped slide-warmers are perfect for cytology, histology or pathology lab.
- Adonized black surface provides contrast
- Digital LED temperature readout
- Ideal for paraffin tissue section mounting
- Covered and uncovered options available
Available in several sizes and can be purchased with or without a cover.
Frequently Bought Together