Normocin™ is an innovative antibiotic formulation designed to prevent cell lines from mycoplasma, bacterial and fungal contaminations. Normocin™ can be routinely added in the cell culture medium in combination with penicillin/streptomycin to broaden the anti-bacterial spectrum.
- Active against both gram+ and gram- bacteria, mycoplasmas and fungi
- Can be used in combination with penicillin/streptomycin
- Non-toxic to the cell line being treated
- One 1 ml vial of Normocin™ is sufficient to treat 500 ml of culture
- One 20 ml bottle of Normocin™ is sufficient to treat 10 liters of culture
- Active concentration not toxic for treated cell lines: 100 μg/ml
- Endotoxin level: <0.5 EU/mg
Normocin™ contains three compounds. Two of these compounds act on gram+ bacteria, gram-acteria, and mycoplasmas by blocking DNA and protein synthesis. The third compound eradicates yeasts and fungi by disrupting ionic exchange through the cell membrane.
Normocin™ should be stored at 4 °C or –20°C.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.