Order Entry
United States
ContactUsLinkComponent
Organic Reference Standard, Anthracene, NSI Lab Solutions
Organic Reference Standard, Anthracene, NSI Lab Solutions
Catalog # NS0076
Supplier:  SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Organic Reference Standard, Anthracene, NSI Lab Solutions
Catalog # NS0076
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  000076
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml in acetone
  • CAS No.:
    120-12-7
  • Description:
    Anthracene
  • Size:
    1 ml
  • Formula:
    C₁₄H₁₀
  • Description:
    Anthracene
  • Cat. No.:
    NS0076
  • Supplier no.:
    000076

Specifications

About this item