VIAFLO Electronic Pipetting Systems, INTEGRA Biosciences AG
Supplier: INTEGRA MS
Certificates
About this item
These VIAFLO electronic pipettes with wide range of GripTips form a perfect pipetting system. The ultra-lightweight, well balanced design of the VIAFLO pipettes feature unsurpassed multichannel pipetting ergonomics, even during prolonged pipetting sessions.
- GripTips snap into place with minimal tip loading effort, providing a secure connection
- Pipettes can be charged directly with the pipette mains adapter
- Versatile user interface and ergonomic design
- Dispensing multiple aliquots of the same volume
The color screen together with the easy to navigate touch wheel user interface promote ease of set up and operation. These pipettes provide a choice of predefined pipetting modes covering typical applications as well as custom multi-step programs for specific tasks.
Ordering information: Power supplies are not included, need to order separately.
GripTips
Product Information
Frequently Bought Together