Order Entry
United States
ContactUsLinkComponent
EDTA disodium salt solution 0.2% (w/v) (synthetic)
EDTA disodium salt solution 0.2% (w/v) (synthetic)
Catalog # IC092820549
Supplier:  MP Biomedicals
CAS Number:  
undefined
EDTA disodium salt solution 0.2% (w/v) (synthetic)
Catalog # IC092820549
Supplier:  MP Biomedicals
Supplier Number:  092820549
CAS Number:  

Specifications

  • Size:
    100 mL
  • Cat. No.:
    IC092820549

Specifications

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672