About this item
BCIP/NBT Liquid Substrate Plus is a stabilized chromogen solution. It has a greater sensitivity and produces little or no background staining compared with other alternatives. It is ready to use solution.
BCIP/NBT [5-bromo-4-chloro-3-indolyl-phosphate p-nitro blue tetrazolium chloride] is in a proprietary buffer and enhancer
The BCIP/NBT Liquid Substrate System offers a convenient, ready-to-use substrate solution for visualizing alkaline phosphatase activity in immunoblotting and immunohistology. Immunohistochemical staining with BCIP/NBT requires organic mounting media and can be counterstained with Nuclear Fast Red or Light Green.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage Conditions:Store product at +2-8 °C
- Form:Liquid
- Protein/Peptide Name:BCIP/NBT Liquid Substrate Plus
- Concentration:BCIP: 0.692 mmol/liter; NBT: 0.734 mmol/liter