ContactUsLinkComponent
PureCol® Type I Collagen Coated Well Plates 48-Well, Advanced BioMatrix
PureCol® Type I Collagen Coated Well Plates 48-Well, Advanced BioMatrix
Catalog # 103700-686
Supplier:  Advanced Biomatrix
PureCol® Type I Collagen Coated Well Plates 48-Well, Advanced BioMatrix
Catalog # 103700-686
Supplier:  Advanced Biomatrix
Supplier Number:  5181-5EA
Specifications
  • No. of wells:
    48-well
  • Description:
    Purecol® Type I Collagen Coated Well Plates
  • Cat. No.:
    103700-686
Specifications
About this item
Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.
  • Coated with PureCol®-the collagen standard for purity
  • Great for 2D cell culture on a native ECM substrate
  • PureCol® is referenced in >2000 publications
PureCol® Collagen Coated, 48-well Plates with a flat bottom have an uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic.
Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.