Orders LinkContactUsLinkComponent
LaMotte® TesTab® Phosphate Test Kit
Phosphate Test Kit, 1 Kit, 100
  470326-584
 :   LaMotte
 :  5892
LaMotte® TesTab® Phosphate Test Kit
  470326-584
 :  LaMotte
 :  
LaMotte® TesTab® Phosphate Test Kit
LaMotte® TesTab® Phosphate Test Kit
Phosphate Test Kit, 1 Kit, 100
 :  
$40.06
Each
Refill for The Comprehensive Water Monitoring Kit 470004-240 and LaMotte® TesTab® Water Investigation Kit 470007-270
  • 100 Tests
  • Range 0, 1, 2, 4 ppm
Each kit contains easy-to-read diagrammed instructions, a color chart, a test factor flash card and all the necessary apparatus and non-hazardous TesTabs® to perform 100 tests
  • Description:
    Phosphate Test Kit
  • Size:
    1 Kit
  • No. of tests:
    100