ContactUsLinkComponent
LaMotte® Total Coliform Test Kit
  470326-586
 :  LaMotte
 :  
undefined
LaMotte® Total Coliform Test Kit
  470326-586
 :  LaMotte
 :  5850
 :  
  • Description:
    Total Coliform Test Kit
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Cat. No.:
    470326-586
  • No. of tests:
    44
An easy-to-use, disposable tube method that indicates the presence of Total Coliform Bacteria.
  • Refill for The Green Basic Water Monitoring Kit 470157-408 and The Green Low Cost Estuary Kit 470146-188
The water sample is placed in a tube containing a reagent tablet and incubated at room temperature for 48 hours. The tube is then examined for positive indicators of coliform bacteria such as color change and formation of gas bubbles. Each kit contains 44 capped tubes each containing a Coliform Test Tablet, and a set of laminated color chart and instruction flashcards.