Order Entry
United States
ContactUsLinkComponent
BSA ELISA Kit, Cygnus Technologies
BSA ELISA Kit, Cygnus Technologies
Catalog # 101120-078
Supplier:  Cygnus
CAS Number:  
BSA ELISA Kit, Cygnus Technologies
Catalog # 101120-078
Supplier:  Cygnus
Supplier Number:  F030
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    BSA ELISA Kit
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Storage Temperature:
    2...8 °C
  • Cat. No.:
    101120-078
  • Supplier no.:
    F030

Specifications

About this item

BSA ELISA kit is intended for quantitating bovine serum albumin (BSA).

This highly sensitive assay for BSA has a detection limit of <250 pg/ml. It can be used to optimize purification processes and in routine quality control of in-process streams, as well as in testing the final product.

Delivery: Kit contains anti-BSA:HRP conjugate, BSA standards set, A-E, TBS wash concentrate, 20X and Anti-BSA coated microtiter strips.