Eppendorf Rotors for Centrifuge 5910 Ri - precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Experience precision in refrigerated centrifugation with Eppendorf Rotors designed for centrifuge 5910 Ri. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorize with adaptable buckets and adaptors, enhancing the versatility of your centrifuge setup. Ideal for temperature-sensitive applications, the Centrifuge 5910 Ri is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated centrifugation experience.
- Catalog No:
- 76204-996
- 76204-998
- 76205-006
- 76205-002
- 76205-004
- 76205-000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001