ContactUsLinkComponent
Flextips™ Swabs, ITW Chemtronics
Flextips™ Swabs, ITW Chemtronics
Catalog # 76240-044
Supplier:  ITW Chemtronics
Flextips™ Swabs, ITW Chemtronics
Catalog # 76240-044
Supplier:  ITW Chemtronics
Supplier Number:  CX50
Specifications
  • Description:
    Flextips™ Swabs
  • Cat. No.:
    76240-044
  • Packaging:
    50 swabs per bag
  • Handle material:
    PP
  • Head material:
    PU foam
Specifications
About this item
Flextips™ Swabs are constructed from medical grade, 100 ppi, polyurethane foam which features fine cell structure, providing excellent particle entrapment characteristics. The foam is thermally bonded to the nylon paddle and the polypropylene swab handle without using adhesives. The tough nylon paddle is very flexible and virtually unbreakable. These swabs are ideal for general cleaning applications.
  • Superior durability
  • Provides gentle, nonabrasive cleaning of sensitive surfaces
  • 100 ppi foam head
  • Traps surface particles while cleaning
  • High solvent capacity; holds solvent well
Fiberless construction does not generate loose fibers or particles. No adhesives or binders are used in the construction. Economical for high quantity applications. Used with clean excess adhesives after gluing, Clean micro-mechanical devices, Remove Flux residues from printed circuit boards and Apply small precise quantities of adhesive or lubricants.