ContactUsLinkComponent
Taklon Flow-Through Luer Lock Brush, Gordon Brush
Taklon Flow-Through Luer Lock Brush, Gordon Brush
  76246-798
Taklon Flow-Through Luer Lock Brush, Gordon Brush
  76246-798
 :  FTWT-18
  • Bristle color:
    White
  • Bristle material:
    Taklon
  • Brush head Ø:
    8 mm (5/16")
  • Description:
    Taklon Flow-Through Luer Lock Brush, 18 Gauge
  • Cat. No.:
    76246-798
  • Trim length:
    ⁷⁄₁₆"
This stainless steel flow-thru needle has a luer lock base, inserted into a nickel-plated ferrule.