Order Entry
United States
ContactUsLinkComponent
Splicing Tool, Splicetronics Solutions
Splicing Tool, Splicetronics Solutions
Catalog # 76251-566
Splicing Tool, Splicetronics Solutions
Catalog # 76251-566
Supplier Number:  ST410

Specifications

  • Description:
    Splice Tool
  • Dimensions:
    20×6×6 cm
  • Cat. No.:
    76251-566
  • Weight:
    0.5 kg

Specifications

About this item

Stapler style splice tool designed specifically for SMT carrier tapes. Used with the splice clip/shim frame and splice tapes of any width. Ensures SMT component reels can be joined quickly and securely.

  • Slider to push splice clips forward
  • Carbon steel
Delivery: It includes clamps to secure reels.