ContactUsLinkComponent
Adhesives Static Mixers, Ellsworth Adhesives
Adhesives Static Mixers, Ellsworth Adhesives
Catalog # 76253-284
Adhesives Static Mixers, Ellsworth Adhesives
Catalog # 76253-284
Supplier Number:  EDS430-24S
Specifications
  • Description:
    Static Mixer Red, 24 Elements
  • Dimensions:
    0.625×0.375"
  • Cat. No.:
    76253-284
Specifications
About this item
Adhesives static mixers are used to automatically mix two part adhesives during application. The various sizes and alternating angled elements ensure consistent and accurate dispensing with products of various viscosities and cure times.
  • Enhance control, produce less waste and save time by eliminating manual mixing
Static Mixer Red is a static disposable mixer with a stepped mixing nozzle outlet and a bell mount cartridge attachment.