Specifications
- Description:Spray Valves, 2x Round Pattern
- Cat. No.:76277-114
- Model:781S-SS-WA
Specifications
About this item
The 781S Series Low Volume Low Pressure (LVLP) spray systems apply consistent coatings of low- to medium-viscosity fluids exactly where needed.
- Consistent area of coverage
- No clogging, dripping or drying out
- No overspray, no mist, no bounce
- Adjustable nozzle air
Microliter to milliliter amounts can be reliably dispensed in round patterns with diameters ranging from 4.3 to 50.8 mm (0.17 to 2.0") and in fan patterns with widths up to 165.1 mm (6.5").