Phillips® Power Bits, Wiha Tools
Supplier: WIHA TOOLS TE
Certificates
About this item
The ideal multi-purpose bits for all around quality and performance.
- Highly resistant to wear and tear for a long tool life
- High grade CVM steel, hardened torsion zone to prevent uneven torque distribution
These are suitable for all applications for both manual and machine Hardness HRC 59-61.
Certifications: DIN 3126 ISO 1173.
Frequently Bought Together