Specifications
- Ø int.:2.54 mm (0.1")
- Description:Disposable Pinch Tube Pen, Natural, PE
- Cat. No.:76251-616
Specifications
About this item
he TS1201 is a hand-held disposable pinch tube pen that provides control for continuous or micro shot applications of low to semi-viscous fluids. Once the dispense cycle is complete, the operator releases the trigger handle and the internal spring forces the shut-off piston to "pinch" the disposable feed path,
preventing further fluid flow. The TS1201 is fitted with a built in flow rate adjustment.
- Disposable "pinch" tube design
- Simple to operate
- Compact size and weight
The only part of the valve making contact with the fluid being dispensed is the disposable pinch tube assembly making the TS1201 ideally suited to dispensing 2-part and CA (Cyanoacrylate) fluids. Black pinch tubes are made available for use with UV adhesives.
The disposable pinch tubes come pre-assembled with a male Luer lock connection for quick and easy tip attachment.