ContactUsLinkComponent
Pneumatic Dispenser, Ultra 850D, EFD
Pneumatic Dispenser, Ultra 850D, EFD
Catalog # 76238-352
Pneumatic Dispenser, Ultra 850D, EFD
Catalog # 76238-352
Supplier Number:  850D
Specifications
  • Description:
    Pneumatic Dispenser, Ultra 850D, Wall Mounted
  • Cat. No.:
    76238-352
Specifications
About this item
Fluid dispenser with analog air and vacuum control. Complete with an introductory supply of dispensing components.
Output pressure 0 to 100 psi for all fluids.