Tip Tinner, Lead Free, R & R Lotion
Catalog # 76241-918
Supplier: R & R Lotion
Specifications
- Description:Tip Tinne, Lead Free, 1.5 oz.
- Environmentally Preferable:
- Cat. No.:76241-918
Specifications
About this item
All the major manufacturers of soldering tips have one thing in common, they all recommend that “Tin Soldering Tips”. I.C. Lead Free Tip Tinner has been formulated to prolong soldering tip life in the harsh environment of no clean and water soluble fluxes.