ContactUsLinkComponent
EV Series Automated Dispensing System, EFD
  76249-670
EV Series Automated Dispensing System, EFD
  76249-670
 :  7360858
  • Description:
    EV Series Automated Dispensing System
  • Dimensions:
    27W×25D×25" H
  • Cat. No.:
    76249-670
EV series automated systems include simple vision for precise fluid dispensing using EFD syringe barrel and valve systems.
  • On-screen preview of the dispensing path facilitates programming
  • Streamlined file import and conversion
  • Wide range of work envelopes
  • Faster cycle and batch times
Specialized vision-guided DispenseMotion™ software and a simple pencil camera make EFD automated systems quick to set up and easy to program. True three-dimensional motion control allows easy programming of dots, lines, circles, arcs and compound arcs.
EV series can work as a standalone system or as a key part of an automated solution and is easily integrated into in-line transfer systems, rotary tables and pallet assembly lines.