Proto® Socket Driver, Testequity LLC TE
Catalog # 76267-918
Supplier: TESTEQUITY LLC TE
Specifications
- Description:Proto® Socket Driver, Six Point
- Length:⅞"
- Size:7/16"
- Cat. No.:76267-918
- Weight:0.05 lbs.
- Finish:Full polish
- Drive size:1/4"
Specifications
About this item
Proto® 6 Point Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials.
The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. This also helps to minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners.
The TorquePlus™ configuration employs a radius-corner design, shifting contact area and stress away from the fastener corners and distributing contact stresses over a large area of the socket, minimizing slippage.
Certifications: Meets ASME B107.110-2012 and SAE AS4984A specifications.