Ground Cord for Workmat, Menda
Catalog # 76242-578
Supplier: Desco
Specifications
- Color:—
- Description:Banana Jack Cord, 10'
- Cat. No.:76242-578
Specifications
About this item
Ground cord for workmat, compatible with mats that have a 10 mm socket snap contain four banana jacks provides for four grounding points for operators, visitors, and/or carts.
- Compact design to fits neatly under bench top - does not use valuable worksurface space
- Allows unit to be permanently installed to workbench
- Extra ring terminal included
Ring terminal at end of ground cord allows easy attachment to electrical ground.
Certifications: Meets grounding standard ANSI/ESD S6.1.