ContactUsLinkComponent
ESD Wash Bottle, R & R Lotion
ESD Wash Bottle, R & R Lotion
Catalog # 76239-634
Supplier:  R & R Lotion
undefined
ESD Wash Bottle, R & R Lotion
Catalog # 76239-634
Supplier:  R & R Lotion
Supplier Number:  WHB-16-ESD
Specifications
  • Capacity:
    16 oz.
  • Description:
    ESD Wash Bottle
  • Cat. No.:
    76239-634
Specifications
About this item
This cleanroom blue static dissipative solvent containers have an average surface resistivity of 10⁹ to 10¹⁰.
  • No chemical reactivity with solvents such as TCE or alcohol
  • No color change or any other physical change noted for any solvents tested
  • Low density ESD safe with closure
  • 90 degree stem molded in one piece for leak proof service
The pH of the deionized water solutions tested was the same before and after a 7-day period.