ContactUsLinkComponent
Connection Validation Soldering Station, CV-5210, OK International
Connection Validation Soldering Station, CV-5210, OK International
Catalog # 76253-714
Connection Validation Soldering Station, CV-5210, OK International
Catalog # 76253-714
Supplier Number:  CV-5332
Specifications
  • Description:
    CV-5210 Connection Validation Soldering Station
  • Environmentally Preferable:
  • Cat. No.:
    76253-714
  • Tip size:
    2.8" Diagonal
  • Power consumption:
    125 W
Specifications
About this item
Metcal soldering stations with SmartHeat® technology. SmartHeat senses the thermal load and instantly delivers Power on Demand directly to the joint.
  • 2.8" color touchscreen with bold graphics
  • Communications Port for process traceability data and firmware graphics
  • Precise tip temperature display with Integrated Net Power Meter and power graph
  • Patented Chip-in-Cartridge technology
  • Closed loop bi-directional communication
The embedded, self-regulating heater delivers the right amount of power based on the requirements of the solder joint. This provides a fast, safe and repeatable process with no calibration required. Stores and records cartridge attributes. Provides traceability information. Protects power supply from non-conforming cartridges. Backwards compatible with MX series power supplies. With password protection.
The intermetallic compound thickness is critical in the formation of a solder joint. Connection Validation (CV) evaluates the quality of the solder joint by calculating the intermetallic compound formation and provides closed loop feedback to the operator. CV provides feedback to the operator via the LED light ring integrated in the hand-piece. It takes the risk associated with the variables involved in the soldering activity and removes the success and failure determination of a good solder joint out of the hands of the operator. CV complements the skill of the operator to judge the quality of a solder joint by introducing an objective method of evaluating solder joint quality. This systemic and objective approach provides repeatability and a measureable standard to the soldering process.